Assay Details
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109938.1 | 9 - 10 | 1638 | 70 | ||
GenBank mRNA | BC079884.1 | - | 9 - 10 | 1583 | 6840050 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Abcb6 | 1200005B17Rik | -5.4kb | Sense | Chr. 1 - 75171640 - 75180283 on NCBI Build GRCm38 |
Downstream | ankyrin repeat and zinc finger domain containing 1 | Ankzf1 |
1300008P06Rik, 2810025E10Rik, D1Ertd161e |
6.1kb | Anti-sense | Chr. 1 - 75192160 - 75199387 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
