Assay Details
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301304.1 | NP_001288233.1 | 1 - 2 | 169 | 77 | |
NR_125731.1 | 1 - 2 | 167 | 77 | |||
GenBank mRNA | AF071070.1 | - | 1 - 2 | 169 | 77 | |
AF071071.1 | - | 1 - 2 | 167 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactomedin-like 3 | Olfml3 |
2810002E22Rik, AI115209, AW550633, HNOEL-iso, ONT3, mONT3 |
-39.2kb | Sense | Chr. 3 - 103735394 - 103738001 on NCBI Build GRCm38 |
Downstream |
DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae) |
Dclre1b |
AI452214, Apollo, SNMIB, mSNM1B |
9.0kb | Sense | Chr. 3 - 103800605 - 103809387 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF071070.1, AF071071.1, NM_001301304.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
