Assay Details
Gene Symbol: E4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF126967.1 | - | 4 - 5 | 629 | 68 | |
AK033285.1 | - | 4 - 5 | 638 | 68 | ||
AK077014.1 | - | 687 | 68 | |||
AK144176.1 | - | 4 - 5 | 583 | 68 | ||
BC005634.1 | - | 4 - 5 | 558 | 68 | ||
BC031757.1 | - | 4 - 5 | 612 | 68 | ||
BC046459.1 | - | 4 - 5 | 696 | 5322519 | 68 | |
BC057011.1 | - | 3 - 4 | 382 | 68 | ||
BC071228.1 | - | 4 - 5 | 639 | 4915902 | 68 | |
X76858.1 | - | 3 - 4 | 422 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | deoxyribonuclease 1-like 2 | Dnase1l2 | 4733401H14Rik | -3.3kb | Sense | Chr. 17 - 24440087 - 24443765 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4833447I15 gene | 4833447I15Rik | 13.7kb | Sense | Chr. 17 - 24461220 - 24470191 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
