Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145569.4 | NP_663544.1 | 8 - 9 | 1206 | 115 | |
GenBank mRNA | AB070266.1 | - | 8 - 9 | 1170 | 115 | |
AK034757.1 | - | 8 - 9 | 1207 | 115 | ||
AK052764.1 | - | 8 - 9 | 1205 | 115 | ||
AK079420.1 | - | 8 - 9 | 1212 | 115 | ||
AK145407.1 | - | 8 - 9 | 1205 | 115 | ||
AK150040.1 | - | 8 - 9 | 1205 | 115 | ||
AK164137.1 | - | 8 - 9 | 1205 | 115 | ||
AK166671.1 | - | 8 - 9 | 1205 | 115 | ||
AK168155.1 | - | 8 - 9 | 1205 | 115 | ||
AK169705.1 | - | 8 - 9 | 1205 | 115 | ||
AK209257.1 | - | 2 - 3 | 269 | 115 | ||
BC003451.1 | - | 8 - 9 | 1146 | 2655444 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
