Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168356.1 | NP_001161828.1 | 8 | 1359 | 112 | |
NM_134253.2 | NP_599014.2 | 10 | 1587 | 112 | ||
GenBank mRNA | AK142472.1 | - | 8 | 1216 | 112 | |
BC005659.1 | - | 10 | 1496 | 3588701 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
