Assay Details
Gene Symbol: Gm15938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BB284599.1 | - | 1 - 2 | 188 | 67 |
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021004.1 | - | 310 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
