Assay Details
Gene Symbol: Slco2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030784.1 | - | 1 | 3320 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slco2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030784.1 | - | 1 | 3320 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |