Assay Details
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155152.1 | - | 1 - 2 | 900 | 97 | |
AK155512.1 | - | 1 - 2 | 894 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
