Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 15 | 2500 | 145 | |
GenBank mRNA | AK151188.1 | - | 15 | 2505 | 145 | |
AK152347.1 | - | 15 | 2505 | 145 | ||
AK152988.1 | - | 15 | 2512 | 145 | ||
AK154745.1 | - | 15 | 2506 | 145 | ||
AK160187.1 | - | 9 | 1604 | 145 | ||
BC016595.1 | - | 15 | 2471 | 3153847 | 145 | |
BC022956.1 | - | 14 | 2604 | 145 | ||
BC024446.1 | - | 6 | 846 | 145 | ||
BC026575.1 | - | 15 | 2482 | 4955015 | 145 | |
S71186.1 | - | 15 | 2500 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
