Assay Details
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008866.1 | - | 1 | 152 | 112 | |
AK141895.1 | - | 1 | 1369 | 112 | ||
AK151977.1 | - | 1 | 898 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
