Assay Details
Gene Symbol: Aldh1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027406.1 | NP_081682.1 | 3 | 376 | 91 | |
GenBank mRNA | BC022112.1 | - | 3 | 376 | 91 | |
BC024055.1 | - | 3 | 333 | 5098789 | 91 | |
BC025939.1 | - | 3 | 358 | 5099360 | 91 | |
BC028817.1 | - | 3 | 323 | 5101815 | 91 | |
BC030722.1 | - | 3 | 360 | 5098671 | 91 | |
BC030723.1 | - | 3 | 292 | 5100007 | 91 | |
BC030727.1 | - | 3 | 362 | 5099026 | 91 | |
BC030730.1 | - | 3 | 376 | 5101835 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
