Assay Details
Gene Symbol: Cd24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009846.2 | NP_033976.1 | 2 | 1599 | 108 | |
GenBank mRNA | AA154829.1 | - | 1 | 197 | 108 | |
AK133869.1 | - | 2 | 1598 | 108 | ||
AK169613.1 | - | 2 | 1596 | 108 | ||
BC023197.1 | - | 1 | 1323 | 108 | ||
BC026702.1 | - | 2 | 1608 | 108 | ||
BC075622.1 | - | 2 | 1579 | 6415159 | 108 | |
M58661.1 | - | 2 | 1587 | 107 | ||
X53825.1 | - | 2 | 1536 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
