Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 18 | 2974 | 121 | |
NM_026653.2 | NP_080929.1 | 17 | 2911 | 121 | ||
GenBank mRNA | AK035084.1 | - | 17 | 2839 | 121 | |
BE981198.1 | - | 1 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
