Assay Details
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177353.1 | NP_001170824.1 | 1 - 2 | 612 | 80 | |
NM_001177354.1 | NP_001170825.1 | 1 - 2 | 612 | 80 | ||
GenBank mRNA | AK133952.1 | - | 1 - 2 | 389 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK133952.1, NM_001177354.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
