Assay Details
Gene Symbol: Bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK160246.1 | - | 1 - 2 | 329 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK160246.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
