Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 15 - 16 | 2111 | 91 | |
GenBank mRNA | AK029009.1 | - | 4 - 5 | 313 | 91 | |
AK030158.1 | - | 7 - 8 | 810 | 91 | ||
L11625.1 | - | 6 - 7 | 649 | 91 | ||
U21301.1 | - | 15 - 16 | 2111 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
