Assay Details
Gene Symbol: Adra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009633.3 | NP_033763.2 | 1 | 1423 | 78 | |
GenBank mRNA | AF332049.1 | - | 1 | 1129 | 78 | |
AF332050.1 | - | 1 | 1129 | 78 | ||
BC066862.1 | - | 1 | 1423 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
