Assay Details
Gene Symbol: Nr0b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011850.2 | NP_035980.1 | 1 - 2 | 579 | 86 | |
GenBank mRNA | AK004930.1 | - | 1 - 2 | 589 | 86 | |
AY360323.1 | - | 1 - 2 | 544 | 86 | ||
AY360324.1 | - | 1 - 2 | 544 | 86 | ||
BC019540.1 | - | 1 - 2 | 579 | 4193866 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
