Assay Details
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018732.3 | NP_061202.3 | 8 - 9 | 1310 | 98 | |
GenBank mRNA | AK148881.1 | - | 7 - 8 | 2019 | 98 | |
CX562753.1 | - | 5 - 6 | 600 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
