Assay Details
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025379.2 | NP_079655.1 | 2 - 3 | 297 | 113 | |
GenBank mRNA | AK002593.1 | - | 2 - 3 | 244 | 113 | |
AK003414.1 | - | 2 - 3 | 248 | 113 | ||
AK132086.1 | - | 2 - 3 | 293 | 113 | ||
BC024350.1 | - | 2 - 3 | 238 | 4987472 | 113 | |
BC089538.1 | - | 2 - 3 | 297 | 6533206 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
