Assay Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042542.2 | NP_001036007.1 | 5 - 6 | 2517 | 63 | |
NM_009651.4 | NP_033781.2 | 5 - 6 | 2513 | 63 | ||
GenBank mRNA | BC130271.1 | - | 5 - 6 | 2445 | 40143193 | 63 |
U07423.1 | - | 5 - 6 | 2486 | 63 | ||
U10341.1 | - | 5 - 6 | 2518 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
