Assay Details
Gene Symbol: Klhl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172565.2 | NP_766153.1 | 1 - 2 | 599 | 108 | |
GenBank mRNA | AK029101.1 | - | 1 - 2 | 600 | 108 | |
AK169910.1 | - | 2 - 3 | 497 | 107 | ||
BC111873.1 | - | 1 - 2 | 595 | 40059437 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK169910.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
