Assay Details
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135172.1 | NP_001128644.1 | 2 - 3 | 663 | 97 | |
NM_175425.4 | NP_780634.2 | 2 - 3 | 306 | 97 | ||
GenBank mRNA | AK078818.1 | - | 2 - 3 | 469 | 97 | |
BC038308.1 | - | 2 - 3 | 306 | 97 | ||
BC090967.1 | - | 2 - 3 | 644 | 30652983 | 97 | |
DQ002400.1 | - | 2 - 3 | 315 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
