Assay Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146176.1 | NP_001139648.1 | 3 - 4 | 486 | 71 | |
NM_008558.2 | NP_032584.2 | 4 - 5 | 513 | 71 | ||
GenBank mRNA | AK080431.1 | - | 3 - 4 | 441 | 71 | |
BC138671.1 | - | 4 - 5 | 369 | 8861692 | 71 | |
BC138672.1 | - | 4 - 5 | 369 | 8861693 | 71 | |
BC145369.1 | - | 3 - 4 | 342 | 9053927 | 71 | |
M63903.1 | - | 4 - 5 | 450 | 71 | ||
S80521.1 | - | 3 - 4 | 182 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
