Assay Details
Gene Symbol: Ceacam14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025957.4 | NP_080233.1 | 2 | 323 | 73 | |
GenBank mRNA | AK005390.1 | - | 2 | 324 | 73 | |
AK005482.1 | - | 2 | 324 | 73 | ||
AK005533.1 | - | 2 | 324 | 73 | ||
BC099412.1 | - | 2 | 253 | 30787387 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
