Assay Details
Gene Symbol: Bcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177410.2 | NP_803129.2 | 1 | 2024 | 59 | |
GenBank mRNA | AK077913.1 | - | 1 | 804 | 59 | |
BC068988.1 | - | 1 | 912 | 1265430 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
