Assay Details
Gene Symbol: Il3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010556.4 | NP_034686.2 | 2 - 3 | 236 | 69 | |
GenBank mRNA | A02046.1 | - | 2 - 3 | 234 | 69 | |
AK153634.1 | - | 2 - 3 | 237 | 69 | ||
BC125552.1 | - | 2 - 3 | 231 | 40130067 | 69 | |
BC125554.1 | - | 2 - 3 | 231 | 40130069 | 69 | |
K01668.1 | - | 2 - 3 | 234 | 69 | ||
K01850.1 | - | 2 - 3 | 234 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
