Assay Details
Gene Symbol: Il13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008355.3 | NP_032381.1 | 1 - 2 | 213 | 79 | |
GenBank mRNA | M23504.1 | - | 1 - 2 | 214 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
- Authors
- Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
- Journal
- J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 184 (2010)
- Product Usage
- -
