Assay Details
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008366.3 | NP_032392.1 | 2 - 3 | 294 | 91 | |
GenBank mRNA | AF065914.1 | - | 2 - 3 | 292 | 91 | |
AF065915.1 | - | 2 - 3 | 292 | 91 | ||
AF065916.1 | - | 2 - 3 | 292 | 91 | ||
AY147902.1 | - | 2 - 3 | 246 | 91 | ||
BC116845.1 | - | 2 - 3 | 292 | 40126164 | 91 | |
BC116873.1 | - | 2 - 3 | 292 | 40126192 | 91 | |
DQ836354.1 | - | 2 - 3 | 186 | 91 | ||
K02292.1 | - | 2 - 3 | 294 | 91 | ||
U41494.1 | - | 2 - 3 | 189 | 91 | ||
U41504.1 | - | 2 - 3 | 204 | 91 | ||
U41505.1 | - | 2 - 3 | 216 | 91 | ||
U41506.1 | - | 2 - 3 | 189 | 91 | ||
X01772.1 | - | 2 - 3 | 293 | 91 | ||
X73040.1 | - | 2 - 3 | 293 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -


Gene Ontology Categories:
