Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967393.1 | XP_027823194.1 | 4 - 5 | 655 | 87 | |
XM_027967394.1 | XP_027823195.1 | 4 - 5 | 655 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
