Assay Details
Gene Symbol: MIDEAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972001.1 | XP_027827802.1 | 11 - 12 | 3238 | 62 | |
XM_027972002.1 | XP_027827803.1 | 11 - 12 | 3644 | 62 | ||
XM_027972003.1 | XP_027827804.1 | 11 - 12 | 3238 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
