Assay Details
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246263.2 | NP_001233192.2 | 1 - 2 | 627 | 76 | |
GenBank mRNA | HQ678172.1 | - | 1 - 2 | 354 | 76 | |
JQ941956.1 | - | 1 - 2 | 627 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
