Assay Details
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009796.1 | NP_001009796.1 | 2 - 3 | 629 | 97 | |
XM_027975567.1 | XP_027831368.1 | 1 - 2 | 586 | 97 | ||
GenBank mRNA | X03883.1 | - | 629 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
