Assay Details
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008726.4 | XP_004008775.1 | 2 - 3 | 464 | 57 | |
XM_004008727.3 | XP_004008776.1 | 2 - 3 | 451 | 57 | ||
XM_012178328.3 | XP_012033718.1 | 2 - 3 | 289 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
