Assay Details
Gene Symbol: PRDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280704.1 | NP_001267633.1 | 4 - 5 | 640 | 65 | |
GenBank mRNA | KC342239.1 | - | 4 - 5 | 640 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
