Assay Details
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177679.1 | NP_001171150.1 | 1 - 2 | 86 | 73 | |
GenBank mRNA | HM012713.1 | - | 1 - 2 | 86 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
