Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006589.3 | XP_004006638.1 | 3 - 4 | 254 | 76 | |
XM_004006590.3 | XP_004006639.1 | 3 - 4 | 254 | 76 | ||
XM_027967603.1 | XP_027823404.1 | 2 - 3 | 196 | 76 | ||
XM_027967604.1 | XP_027823405.1 | 2 - 3 | 196 | 76 | ||
GenBank mRNA | EU370541.1 | - | 1 - 2 | 151 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
