Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006081.3 | XP_004006130.1 | 6 - 7 | 752 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
