Assay Details
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010430.4 | XP_004010479.2 | 1 - 2 | 310 | 65 | |
XM_012181160.3 | XP_012036550.2 | 1 - 2 | 310 | 65 | ||
XM_012181161.3 | XP_012036551.2 | 1 - 2 | 310 | 65 | ||
XM_012181162.3 | XP_012036552.2 | 1 - 2 | 310 | 65 | ||
XM_027971655.1 | XP_027827456.1 | 1 - 2 | 310 | 65 |
