Assay Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002680.4 | XP_004002729.1 | 7 - 8 | 1503 | 72 | |
XM_012184689.3 | XP_012040079.1 | 8 - 9 | 1372 | 72 | ||
XM_015092335.2 | XP_014947821.1 | 7 - 8 | 1507 | 72 | ||
XM_015092338.2 | XP_014947824.1 | 7 - 8 | 1507 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
