Assay Details
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306110.1 | NP_001293039.1 | 7 - 8 | 1203 | 67 | |
XM_027975558.1 | XP_027831359.1 | 6 - 7 | 3739 | 67 | ||
XM_027975559.1 | XP_027831360.1 | 7 - 8 | 3120 | 67 | ||
XM_027975560.1 | XP_027831361.1 | 6 - 7 | 3671 | 67 | ||
GenBank mRNA | KJ159212.1 | - | 7 - 8 | 1203 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
