Assay Details
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018930.4 | XP_004018979.2 | 8 - 9 | 1280 | 58 | |
XM_012100921.3 | XP_011956311.2 | 8 - 9 | 1284 | 58 | ||
XM_012100922.3 | XP_011956312.2 | 8 - 9 | 1282 | 58 | ||
XM_027958809.1 | XP_027814610.1 | 8 - 9 | 1295 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
