Assay Details
Gene Symbol: DARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161887.1 | NP_001155359.1 | 7 - 8 | 634 | 90 | |
GenBank mRNA | FJ969422.1 | - | 7 - 8 | 634 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
