Assay Details
Gene Symbol: CIBAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184137.3 | XP_012039527.2 | 3 - 4 | 429 | 102 | |
XM_012184138.3 | XP_012039528.2 | 3 - 4 | 429 | 102 | ||
XM_012184140.3 | XP_012039530.2 | 3 - 4 | 429 | 102 | ||
XM_012184142.3 | XP_012039532.2 | 3 - 4 | 504 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
