Assay Details
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005762.4 | XP_004005811.2 | 5 - 6 | 423 | 67 | |
XM_015094269.2 | XP_014949755.2 | 5 - 6 | 431 | 67 | ||
XM_015094270.2 | XP_014949756.1 | 6 - 7 | 488 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
