Assay Details
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008474.3 | XP_004008523.1 | 3 - 4 | 630 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008474.3 | XP_004008523.1 | 3 - 4 | 630 | 90 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |