Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015105113.2 | XP_014960599.2 | 6 - 7 | 1371 | 64 | |
XM_027962884.1 | XP_027818685.1 | 6 - 7 | 1363 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
