Assay Details
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017049.4 | XP_004017098.2 | 1 - 2 | 397 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017049.4 | XP_004017098.2 | 1 - 2 | 397 | 114 |
Set Membership: |
![]() ![]() ![]() |