Assay Details
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005829.4 | XP_004005878.2 | 3 - 4 | 551 | 93 | |
XM_027966687.1 | XP_027822488.1 | 3 - 4 | 551 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
