Assay Details
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009452.1 | NP_001009452.1 | 2 - 3 | 267 | 97 | |
GenBank mRNA | AF095782.1 | - | 1 - 2 | 180 | 97 | |
AJ417904.1 | - | 2 - 3 | 267 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
